Tlačiť
Gas Washing Bottle 250 ml, Head with VitraPOR® Filter Disc, Standard Joint NS 29/32
Dodávateľ: ROBU GLASFILTER
40252 40254 40250 40251 40253
ROBU40252EA 106.7 EUR
ROBU40252 ROBU40254 ROBU40250 ROBU40251 ROBU40253
Gas Washing Bottle 250 ml, Head with VitraPOR® Filter Disc, Standard Joint NS 29/32
Láhve Promývačka plynů
ROBU® gas washing bottle head with integrated VitraPOR® filter disc and standard ground joint ensures efficient gas-liquid contact and uniform dispersion in absorption processes.
  • Borosilicate glass
  • High chemical and thermal resistance
  • Compatible with standard ground joint heads
  • Robust and reusable
  • Ideal for use with VitraPOR® heads
  • Clear visibility of contents
  • NSH 29/32
The ROBU Gas Washing Bottle Head with VitraPOR Filter Disc and Standard Joint is designed for efficient gas scrubbing in laboratory setups. The integrated filter disc ensures fine dispersion of gases into the washing liquid, maximising surface contact and absorption efficiency. Made from high-quality borosilicate glass, it offers excellent chemical resistance and durability. The standardized joint ensures a secure, leak-free fit with compatible gas washing bottles. Ideal for removing impurities, neutralizing aggressive gases, or humidifying gas streams, the ROBU head component guarantees reliable performance in chemical processing and analytical applications.
Certifikácie:
In compliance with ISO 4796-80 and ASTM E128-99 standards.