Tlačiť
HexArmor® Hercules™ NSR 3041 Safety Gloves, 60983 Series
Dodávateľ: UVEX
6098307 6098308 6098309 6098310 6098311
UVEX6098307EA 0 EUR
UVEX6098307 UVEX6098308 UVEX6098309 UVEX6098310 UVEX6098311
HexArmor® Hercules™ NSR 3041 Safety Gloves, 60983 Series
Rukavice
The Hercules™ NSR 3041 glove is engineered to provide the highest levels of needle and cut protection available on the market.
  • 360° degree cut and needle protection
  • Form-fitting design for increased dexterity and comfort
  • Launderable
This glove offers full coverage over the entire hand for the most harsh applications and was tested using 25-gauge needles. SuperFabric® material protects against hazards through a configuration of tiny guard plates layered onto performance fabrics. Gloves feature three layers of SuperFabric® material on the palm (interior layer) and back-of-hand and two layers on the finger sidewalls. Reinforced fingertips and back-of-hand knuckle padding provide increased protection for incidental bumps and impacts. A specialized silicone gripping surface on the palm provides enhanced durability and grip.