Tlačiť
Cubis® II Manual Mass Comparators
Dodávateľ: Sartorius Balances
MCA1004MC MCA1005MC MCA106MC MCA206MC MCA36MC MCA605MC MCA66MC
SARBMCA1004MCEA 43750.5 EUR
SARBMCA1004MC SARBMCA1005MC SARBMCA106MC SARBMCA206MC SARBMCA36MC SARBMCA605MC SARBMCA66MC
Cubis® II Manual Mass Comparators
Váhy
The Cubis® II Manual mass comparators are the latest addition to the portfolio of the Sartorius Mass Comparators. They bring mass determination to a new level by uniting metrological weighing competence and newest hardware technology with an integrated control of workflows in line with the recommendations of the International  Organization of Legal Metrology (OIML) and are compliant with ASTM.
  • Next generation monolithic weighing system guarantees the highest level of accuracy
  • Reduced influence of air drafts and ease of use thanks to optimised weighing chamber size and automated draft shields
  • Environment adaptation capability and fast stabilisation time ensured by novel engineering solutions
  • Integrated workflows for error-free mass calibration using the MassCal App
  • Integrated climate sensor further support measurement accuracy
  • Traceable cleaning process guidance with the help of the Cleaning QApp
Cubis® II Manual mass comparators are specifically designed for determining the conventional value of mass and its standard uncertainty without compromising on performance. The latest generation of monolithic weighing systems offer fast stabilisation times and low repeatability even under F class conditions.
Certifikácie:
EN 61326-1:2013, EN IEC 61326:2021,
EN IEC 63000:2018.