Tlačiť
Human PDGF BB ELISA Kits
Dodávateľ: Abcam
AB184860-96 AB184860-10X96
ABCAAB184860-96EA 1093.46 EUR
ABCAAB184860-96 ABCAAB184860-10X96
Human PDGF BB ELISA Kits
Test ELISY
Human PDGF BB ELISA Kit is a single-wash 90-min Simplestep used to quantify Human PDGF BB with a sensitivity of 2,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.