Mouse Erythropoietin ELISA Kits (EPO)
Dodávateľ: Abcam
Mouse Erythropoietin ELISA kit (EPO) is a single-wash 90-min Simplestep used to quantify Mouse Erythropoietin (EPO) with a sensitivity of 2.35 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.