Tlačiť
Mouse TNF alpha ELISA Kit
Dodávateľ: Abcam
AB208348-96 AB208348-1X384
ABCAAB208348-96EA 1079.02 EUR
ABCAAB208348-96 ABCAAB208348-1X384
Mouse TNF alpha ELISA Kit
Test ELISY
Mouse TNF alpha ELISA kit is a single-wash, 90-minute SimpleStep ELISA® designed to quantify mouse TNF alpha with a sensitivity of 9,1 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs - 384-well format for higher throughput - Broad sample compatibility - works with serum, plasma (heparin, citrate, EDTA), and cell culture supernatants.