Tlačiť
Mouse CSF-1-R ELISA Kit
Dodávateľ: Abcam
AB240681-1X96 AB240681-1X384
ABCAAB240681-1X96EA 1114.72 EUR
ABCAAB240681-1X96 ABCAAB240681-1X384
Mouse CSF-1-R ELISA Kit
Test ELISY
Mouse CSF-1-R ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse CSF-1-R with a sensitivity of 0,04 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.