Tlačiť
Human IL-10 ELISA Kit (Interleukin-10)
Dodávateľ: Abcam
AB185986-96 AB185986-1X384
ABCAAB185986-96EA 1114.72 EUR
ABCAAB185986-96 ABCAAB185986-1X384
Human IL-10 ELISA Kit (Interleukin-10)
Test ELISY
Human IL-10 ELISA kit (Interleukin-10) is a single-wash, 90-minute SimpleStep ELISA® assay designed for the quantification of human IL-10 with a sensitivity of 1,4 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput - Broad sample compatibility - works with serum, plasma, and cell culture supernatants.