Tlačiť
Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular
Dodávateľ: Abcam
AB320791-10X96 AB320791-1X384 AB320791-96TST
ABCAAB320791-10X96EA 9324.51 EUR
ABCAAB320791-10X96 ABCAAB320791-1X384 ABCAAB320791-96TST
Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular
Test ELISY
Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular is a single-wash 90-min Simplestep used to quantify Human Apolipoprotein CI/Apo-CI with a sensitivity of 14,44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.