Tlačiť
HexArmor® Protective Sleeves, 60982 Series
Dodávateľ: UVEX
6098201 6098202 6098203 6098204 6098205 6098206
UVEX6098201EA 0 EUR
UVEX6098201 UVEX6098202 UVEX6098203 UVEX6098204 UVEX6098205 UVEX6098206
HexArmor® Protective Sleeves, 60982 Series
Rukavice
Industrial puncture protection for the forearm with spandex wrist insert, thumbhole, and snap fasteners.
  • High cut and needle resistance
  • Won’t fall down like knit sleeves
  • Launderable
Two layers of SuperFabric® material protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. Lightweight design is cooler than knit or leather alternatives.