Tlačiť
Anti-SRP1 Rabbit Monoclonal Antibody [clone: EPR26346-53] (Alexa Fluor® 647)
Dodávateľ: ABCAM
AB316342-10UL AB316342-100UL
ABCAAB316342-10ULEA 271.56 EUR
ABCAAB316342-10UL ABCAAB316342-100UL
Anti-SRP1 Rabbit Monoclonal Antibody [clone: EPR26346-53] (Alexa Fluor® 647)
Protilátky
Alexa Fluor® 647 Rabbit monoclonal [EPR26346-53] to SRP1.
Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SRP1
Clonality: Monoclonal
Clone: EPR26346-53
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human