Tlačiť
Anti-Streptococcus mutans Cell surface antigen I/II Rabbit Monoclonal Antibody [clone: EPR28235-11]
Dodávateľ: ABCAM
AB316006-20UL AB316006-100UL AB316006-1ML
ABCAAB316006-20ULEA 314.48 EUR
ABCAAB316006-20UL ABCAAB316006-100UL ABCAAB316006-1ML
Anti-Streptococcus mutans Cell surface antigen I/II Rabbit Monoclonal Antibody [clone: EPR28235-11]
Protilátky
Rabbit monoclonal [EPR28235-11] to Streptococcus mutans Cell surface antigen I/II.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Streptococcus mutans Cell surface antigen I/II
Clonality: Monoclonal
Clone: EPR28235-11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: STREP