Tlačiť
Anti-IL-23 Rabbit Monoclonal Antibody [clone: EPR26364-40]
Dodávateľ: ABCAM
AB314486-20UL AB314486-100UL AB314486-1ML
ABCAAB314486-20ULEA 278.78 EUR
ABCAAB314486-20UL ABCAAB314486-100UL ABCAAB314486-1ML
Anti-IL-23 Rabbit Monoclonal Antibody [clone: EPR26364-40]
Protilátky
Rabbit monoclonal [EPR26364-40] to IL-23.
Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IL-23
Clonality: Monoclonal
Clone: EPR26364-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human