Tlačiť

Anti-Human respiratory syncytial virus A Nucleoprotein Rabbit Monoclonal Antibody [clone: EPR28656-26]

Dodávateľ: ABCAM
AB316207-20UL AB316207-100UL AB316207-1ML
ABCAAB316207-20ULEA 278.78 EUR
ABCAAB316207-20UL ABCAAB316207-100UL ABCAAB316207-1ML
Anti-Human respiratory syncytial virus A Nucleoprotein Rabbit Monoclonal Antibody [clone: EPR28656-26]
Protilátky
Rabbit monoclonal [EPR28656-26] to Human respiratory syncytial virus A Nucleoprotein.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Human respiratory syncytial virus A Nucleoprotein
Clonality: Monoclonal
Clone: EPR28656-26
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR