Tlačiť
Anti-Complement Factor B (Ba fragment) Rabbit Monoclonal Antibody [clone: EPR24245-52]
Dodávateľ: ABCAM
AB316819-1MG AB316819-100UG
ABCAAB316819-1MGEA 10517.45 EUR
ABCAAB316819-1MG ABCAAB316819-100UG
Anti-Complement Factor B (Ba fragment) Rabbit Monoclonal Antibody [clone: EPR24245-52]
Protilátky
Rabbit monoclonal [EPR24245-52] to Complement Factor B (Ba fragment) - BSA and Azide free (Detector).
Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Complement Factor B (Ba fragment)
Clonality: Monoclonal
Clone: EPR24245-52
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human