Tlačiť

Anti-Streptococcus mutans Cell surface antigen I/II Rabbit Monoclonal Antibody [clone: EPR28235-11]

Dodávateľ: ABCAM
AB316007-1MG AB316007-100UG
ABCAAB316007-1MGEA 10467.71 EUR
ABCAAB316007-1MG ABCAAB316007-100UG
Anti-Streptococcus mutans Cell surface antigen I/II Rabbit Monoclonal Antibody [clone: EPR28235-11]
Protilátky
Rabbit monoclonal [EPR28235-11] to Streptococcus mutans Cell surface antigen I/II - BSA and Azide free.

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Streptococcus mutans Cell surface antigen I/II
Clonality: Monoclonal
Clone: EPR28235-11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptococcus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR