Tlačiť

Anti-POLD1 Rabbit Monoclonal Antibody [clone: EPR15118]

Dodávateľ: ABCAM
AB186407-100 AB186407-1ML
ABCAAB186407-100EA 757.72 EUR
ABCAAB186407-100 ABCAAB186407-1ML
Anti-POLD1 Rabbit Monoclonal Antibody [clone: EPR15118]
Protilátky
Rabbit monoclonal [EPR15118] to POLD1.

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: POLD1
Clonality: Monoclonal
Clone: EPR15118
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR