Tlačiť
Anti-TEM8/ATR Rabbit Monoclonal Antibody [clone: EPNCI-R173-37]
Dodávateľ: ABCAM
AB241067-100 AB241067-1ML
ABCAAB241067-100EA 893.3 EUR
ABCAAB241067-100 ABCAAB241067-1ML
Anti-TEM8/ATR Rabbit Monoclonal Antibody [clone: EPNCI-R173-37]
Protilátky
Rabbit monoclonal [EPNCI-R173-37] to TEM8/ATR.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TEM8/ATR
Clonality: Monoclonal
Clone: EPNCI-R173-37
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse