Tlačiť

Anti-Histone H2B (phospho T129) Rabbit Monoclonal Antibody [clone: EPR18095]

Dodávateľ: ABCAM
AB188292-100 AB188292-1ML
ABCAAB188292-100EA 736.06 EUR
ABCAAB188292-100 ABCAAB188292-1ML
Anti-Histone H2B (phospho T129) Rabbit Monoclonal Antibody [clone: EPR18095]
Protilátky
Rabbit monoclonal [EPR18095] to Histone H2B (phospho T129) - ChIP Grade.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EPR18095
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR