Tlačiť

Anti-IDH2 Rat Monoclonal Antibody [clone: MMab-1]

Dodávateľ: ABCAM
AB264061-100 AB264061-1
ABCAAB264061-100EA 1164.86 EUR
ABCAAB264061-100 ABCAAB264061-1
Anti-IDH2 Rat Monoclonal Antibody [clone: MMab-1]
Protilátky
Rat monoclonal [MMab-1] to IDH2 (mutated R172M) - BSA and Azide free.

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IDH2
Clonality: Monoclonal
Clone: MMab-1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR