Tlačiť

Anti-IgG Goat Polyclonal Antibody (Gold, 40nm)

Dodávateľ: ABCAM
AB270694-200 AB270694-1
ABCAAB270694-200EA 243.08 EUR
ABCAAB270694-200 ABCAAB270694-1
Anti-IgG Goat Polyclonal Antibody (Gold, 40nm)
Protilátky
Goat Anti-Rabbit IgG (40nm Gold, 10 OD).

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, 40nm
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR