Tlačiť

Anti-Histone H2A+Histone H4 (symmetric di methyl R3) Rabbit Monoclonal Antibody [clone: EPR27303-14]

Dodávateľ: ABCAM
AB309354-100UL AB309354-20UL AB309354-1ML
ABCAAB309354-100ULEA 793.42 EUR
ABCAAB309354-100UL ABCAAB309354-20UL ABCAAB309354-1ML
Anti-Histone H2A+Histone H4 (symmetric di methyl R3) Rabbit Monoclonal Antibody [clone: EPR27303-14]
Protilátky
Rabbit monoclonal [EPR27303-14] to Histone H2A (symmetric di methyl R3) + Histone H4 (symmetric di methyl R3).

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H2A+Histone H4
Clonality: Monoclonal
Clone: EPR27303-14
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR