Tlačiť

Anti-MED6 Rabbit Monoclonal Antibody [clone: EPR12107]

Dodávateľ: ABCAM
AB249702-1 AB249702-100
ABCAAB249702-1EA 10467.71 EUR
ABCAAB249702-1 ABCAAB249702-100
Anti-MED6 Rabbit Monoclonal Antibody [clone: EPR12107]
Protilátky
Rabbit monoclonal [EPR12107] to MED6 - BSA and Azide free.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MED6
Clonality: Monoclonal
Clone: EPR12107
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR