Tlačiť
Anti-PAP Rabbit Monoclonal Antibody [clone: EPR22614-226]
Dodávateľ: ABCAM
AB269387-1 AB269387-100
ABCAAB269387-1EA 10646.21 EUR
ABCAAB269387-1 ABCAAB269387-100
Anti-PAP Rabbit Monoclonal Antibody [clone: EPR22614-226]
Protilátky
Rabbit monoclonal [EPR22614-226] to PAP - BSA and Azide free (Detector).
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PAP
Clonality: Monoclonal
Clone: EPR22614-226
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human