Tlačiť
Anti-pro Caspase-1 + p10 + p12 Rabbit Monoclonal Antibody [clone: EPR16883]
Dodávateľ: ABCAM
AB238972-100 AB238972-1
ABCAAB238972-100EA 1164.86 EUR
ABCAAB238972-100 ABCAAB238972-1
Anti-pro Caspase-1 + p10 + p12 Rabbit Monoclonal Antibody [clone: EPR16883]
Protilátky
Rabbit monoclonal [EPR16883] to pro Caspase-1 + p10 + p12 - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: pro Caspase-1 + p10 + p12
Clonality: Monoclonal
Clone: EPR16883
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human