Tlačiť
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61]
Dodávateľ: ABCAM
AB183179-100 AB183179-1
ABCAAB183179-100EA 1164.86 EUR
ABCAAB183179-100 ABCAAB183179-1
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61]
Protilátky
Rabbit monoclonal [E61] to pro Caspase-3 - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: pro Caspase-3
Clonality: Monoclonal
Clone: E61
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human