Tlačiť

Anti-PKC mu/PKD Rabbit Monoclonal Antibody [clone: EP1493Y]

Dodávateľ: ABCAM
AB217484-1 AB217484-100
ABCAAB217484-1EA 0 EUR
ABCAAB217484-1 ABCAAB217484-100
Anti-PKC mu/PKD Rabbit Monoclonal Antibody [clone: EP1493Y]
Protilátky
Rabbit monoclonal [EP1493Y] to PKC mu/PKD - BSA and Azide free.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PKC mu/PKD
Clonality: Monoclonal
Clone: EP1493Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR