Tlačiť

Anti-EEF2/Elongation factor 2 Rabbit Monoclonal Antibody [clone: EP880Y]

Dodávateľ: ABCAM
AB247388-100 AB247388-1
ABCAAB247388-100EA 1164.86 EUR
ABCAAB247388-100 ABCAAB247388-1
Anti-EEF2/Elongation factor 2 Rabbit Monoclonal Antibody [clone: EP880Y]
Protilátky
Rabbit monoclonal [EP880Y] to EEF2/Elongation factor 2 - BSA and Azide free.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: EEF2/Elongation factor 2
Clonality: Monoclonal
Clone: EP880Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR