Tlačiť
Anti-pro-EGF Rabbit Monoclonal Antibody [clone: EPR14622]
Dodávateľ: ABCAM
AB206106-100 AB206106-1ML
ABCAAB206106-100EA 807.46 EUR
ABCAAB206106-100 ABCAAB206106-1ML
Anti-pro-EGF Rabbit Monoclonal Antibody [clone: EPR14622]
Protilátky
Rabbit monoclonal [EPR14622] to pro-EGF.
Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: pro-EGF
Clonality: Monoclonal
Clone: EPR14622
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human