Tlačiť
Anti-Histone H3 (mono methyl K14) Rabbit Monoclonal Antibody [clone: EPR17708-235]
Dodávateľ: ABCAM
AB251366-100 AB251366-1
ABCAAB251366-100EA 1164.86 EUR
ABCAAB251366-100 ABCAAB251366-1
Anti-Histone H3 (mono methyl K14) Rabbit Monoclonal Antibody [clone: EPR17708-235]
Protilátky
Rabbit monoclonal [EPR17708-235] to Histone H3 (mono methyl K14) - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR17708-235
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse