Tlačiť
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61] (Alexa Fluor® 488)
Dodávateľ: ABCAM
AB307610-100 AB307610-10
ABCAAB307610-100EA 936.22 EUR
ABCAAB307610-100 ABCAAB307610-10
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61] (Alexa Fluor® 488)
Protilátky
Alexa Fluor® 488 Rabbit monoclonal [E61] to pro Caspase-3.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: pro Caspase-3
Clonality: Monoclonal
Clone: E61
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human