Tlačiť
Anti-Pro-Collagen III alpha 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1028]
Dodávateľ: ABCAM
AB283725-100 AB283725-1
ABCAAB283725-100EA 1164.86 EUR
ABCAAB283725-100 ABCAAB283725-1
Anti-Pro-Collagen III alpha 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1028]
Protilátky
Rabbit recombinant multiclonal [RM1028] to Pro-Collagen III alpha 1 - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Pro-Collagen III alpha 1
Clonality: Recombinant Multiclonal
Clone: RM1028
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat