Tlačiť

Anti-Myc tag Human Monoclonal Antibody [clone: Hyper-myc]

Dodávateľ: ABCAM
AB289981-100 AB289981-1MG AB289981-20UG
ABCAAB289981-100EA 543.12 EUR
ABCAAB289981-100 ABCAAB289981-1MG ABCAAB289981-20UG
Anti-Myc tag Human Monoclonal Antibody [clone: Hyper-myc]
Protilátky
Human monoclonal [Hyper-myc] to Myc tag.

Type: Primary
Antigen: Myc tag
Clonality: Monoclonal
Clone: Hyper-myc
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR