Tlačiť

Anti-AKT1 (phospho S124 + S124) Rabbit Monoclonal Antibody [clone: EPR17680]

Dodávateľ: ABCAM
AB183556-100 AB183556-20UL AB183556-1ML
ABCAAB183556-100EA 543.12 EUR
ABCAAB183556-100 ABCAAB183556-20UL ABCAAB183556-1ML
Anti-AKT1 (phospho S124 + S124) Rabbit Monoclonal Antibody [clone: EPR17680]
Protilátky
Rabbit monoclonal [EPR17680] to AKT1 (phospho S124).

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: EPR17680
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR