Tlačiť

Anti-AIRE Rat Monoclonal Antibody [clone: 5H12]

Dodávateľ: ABCAM
AB243169-100 AB243169-1MG
ABCAAB243169-100EA 821.9 EUR
ABCAAB243169-100 ABCAAB243169-1MG
Anti-AIRE Rat Monoclonal Antibody [clone: 5H12]
Protilátky
Rat monoclonal [5H12] to AIRE.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AIRE
Clonality: Monoclonal
Clone: 5H12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR