Tlačiť

Anti-Tafazzin / TAZ Rabbit Monoclonal Antibody [clone: EPR26956-58]

Dodávateľ: ABCAM
AB307148-100 AB307148-20UL AB307148-1ML
ABCAAB307148-100EA 793.42 EUR
ABCAAB307148-100 ABCAAB307148-20UL ABCAAB307148-1ML
Anti-Tafazzin / TAZ Rabbit Monoclonal Antibody [clone: EPR26956-58]
Protilátky
Rabbit monoclonal [EPR26956-58] to Tafazzin / TAZ.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tafazzin / TAZ
Clonality: Monoclonal
Clone: EPR26956-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR