Tlačiť

Anti-CD3 zeta (phospho Y83) Rabbit Monoclonal Antibody [clone: EP776(2)Y]

Dodávateľ: ABCAM
AB238955-100 AB238955-1
ABCAAB238955-100EA 1164.86 EUR
ABCAAB238955-100 ABCAAB238955-1
Anti-CD3 zeta (phospho Y83) Rabbit Monoclonal Antibody [clone: EP776(2)Y]
Rabbit monoclonal [EP776(2)Y] to CD3 zeta (phospho Y83) - BSA and Azide free.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD3 zeta
Clonality: Monoclonal
Clone: EP776(2)Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR