Tlačiť

Anti-OB Cadherin Rabbit Polyclonal Antibody

Dodávateľ: ABCAM
AB151302-100 AB151302-25
ABCAAB151302-100EA 757.72 EUR
ABCAAB151302-100 ABCAAB151302-25
Anti-OB Cadherin Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal to OB Cadherin.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: OB Cadherin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR