Tlačiť

Anti-Glutathione Peroxidase 4 Rat Monoclonal Antibody [clone: 1B4]

Dodávateľ: ABCAM
AB255789-1 AB255789-100
ABCAAB255789-1EA 10467.71 EUR
ABCAAB255789-1 ABCAAB255789-100
Anti-Glutathione Peroxidase 4 Rat Monoclonal Antibody [clone: 1B4]
Protilátky
Rat monoclonal [1B4] to Glutathione Peroxidase 4 - BSA and Azide free.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glutathione Peroxidase 4
Clonality: Monoclonal
Clone: 1B4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR