Tlačiť
Anti-Sts1 Rabbit Monoclonal Antibody [clone: EPR16980]
Dodávateľ: ABCAM
AB251234-1 AB251234-100
ABCAAB251234-1EA 10467.71 EUR
ABCAAB251234-1 ABCAAB251234-100
Anti-Sts1 Rabbit Monoclonal Antibody [clone: EPR16980]
Protilátky
Rabbit monoclonal [EPR16980] to Sts1 - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Sts1
Clonality: Monoclonal
Clone: EPR16980
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human