Tlačiť

Anti-MCT 8 Rabbit Monoclonal Antibody [clone: EPR26257-174]

Dodávateľ: ABCAM
AB302707-1 AB302707-100
ABCAAB302707-1EA 10467.71 EUR
ABCAAB302707-1 ABCAAB302707-100
Anti-MCT 8 Rabbit Monoclonal Antibody [clone: EPR26257-174]
Protilátky
Rabbit monoclonal [EPR26257-174] to MCT 8.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MCT 8
Clonality: Monoclonal
Clone: EPR26257-174
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR