Tlačiť

Anti-Myc tag Mouse Monoclonal Antibody [clone: Hyper-myc]

Dodávateľ: ABCAM
AB289980-100 AB289980-20UL AB289980-1ML
ABCAAB289980-100EA 543.12 EUR
ABCAAB289980-100 ABCAAB289980-20UL ABCAAB289980-1ML
Anti-Myc tag Mouse Monoclonal Antibody [clone: Hyper-myc]
Protilátky
Mouse monoclonal [Hyper-myc] to Myc tag - Chimeric.

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Myc tag
Clonality: Monoclonal
Clone: Hyper-myc
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR