Tlačiť
Anti-Lamin B1 Rabbit Monoclonal Antibody [clone: EPR22165-121]
Dodávateľ: ABCAM
AB239399-100 AB239399-1
ABCAAB239399-100EA 1164.86 EUR
ABCAAB239399-100 ABCAAB239399-1
Anti-Lamin B1 Rabbit Monoclonal Antibody [clone: EPR22165-121]
Protilátky
Rabbit monoclonal [EPR22165-121] to Lamin B1 - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Lamin B1
Clonality: Monoclonal
Clone: EPR22165-121
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat