Tlačiť
Anti-Histone H2B (testis specific) Rabbit Monoclonal Antibody [clone: EPR12881]
Dodávateľ: ABCAM
AB250047-100 AB250047-1
ABCAAB250047-100EA 1164.86 EUR
ABCAAB250047-100 ABCAAB250047-1
Anti-Histone H2B (testis specific) Rabbit Monoclonal Antibody [clone: EPR12881]
Protilátky
Rabbit monoclonal [EPR12881] to Histone H2B (testis specific) - BSA and Azide free.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H2B (testis specific)
Clonality: Monoclonal
Clone: EPR12881
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse