Tlačiť

Anti-ENO1 Rabbit Monoclonal Antibody [clone: EPR19758]

Dodávateľ: ABCAM
AB227978-100 AB227978-1ML
ABCAAB227978-100EA 757.72 EUR
ABCAAB227978-100 ABCAAB227978-1ML
Anti-ENO1 Rabbit Monoclonal Antibody [clone: EPR19758]
Protilátky
Rabbit monoclonal [EPR19758] to ENO1.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ENO1
Clonality: Monoclonal
Clone: EPR19758
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR