Tlačiť

Anti-AKT1 + AKT2 (phospho S473) Rabbit Monoclonal Antibody [clone: EPR18405] (Alexa Fluor® 488)

Dodávateľ: ABCAM
AB312893-10UL AB312893-100UL
ABCAAB312893-10ULEA 271.56 EUR
ABCAAB312893-10UL ABCAAB312893-100UL
Anti-AKT1 + AKT2 (phospho S473) Rabbit Monoclonal Antibody [clone: EPR18405] (Alexa Fluor® 488)
Protilátky
Alexa Fluor® 488 Rabbit monoclonal [EPR18405] to AKT1 + AKT2.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AKT1 + AKT2
Clonality: Monoclonal
Clone: EPR18405
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR