Tlačiť
Anti-Poliovirus Receptor/PVR Rabbit Monoclonal Antibody [clone: 305] (FITC)
Dodávateľ: ABCAM
AB275717-25 AB275717-100
ABCAAB275717-25EA 278.78 EUR
ABCAAB275717-25 ABCAAB275717-100
Anti-Poliovirus Receptor/PVR Rabbit Monoclonal Antibody [clone: 305] (FITC)
Protilátky
FITC Rabbit monoclonal [305] to Poliovirus Receptor/PVR.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Poliovirus Receptor/PVR
Clonality: Monoclonal
Clone: 305
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse