Tlačiť

Anti-KAT1/HAT1 Rabbit Monoclonal Antibody [clone: EPR18775]

Dodávateľ: ABCAM
AB194296-100 AB194296-1ML
ABCAAB194296-100EA 921.78 EUR
ABCAAB194296-100 ABCAAB194296-1ML
Anti-KAT1/HAT1 Rabbit Monoclonal Antibody [clone: EPR18775]
Protilátky
Rabbit monoclonal [EPR18775] to KAT1/HAT1.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: KAT1 / HAT1
Clonality: Monoclonal
Clone: EPR18775
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR