Tlačiť
Anti-Histone H2B (testis specific) Rabbit Monoclonal Antibody [clone: EPR12881]
Dodávateľ: ABCAM
AB178426-100 AB178426-1ML
ABCAAB178426-100EA 793.42 EUR
ABCAAB178426-100 ABCAAB178426-1ML
Anti-Histone H2B (testis specific) Rabbit Monoclonal Antibody [clone: EPR12881]
Protilátky
Rabbit monoclonal [EPR12881] to Histone H2B (testis specific).
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H2B (testis specific)
Clonality: Monoclonal
Clone: EPR12881
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse