Tlačiť

Anti-KAT7/Hbo1/MYST2 Rabbit Monoclonal Antibody [clone: EPR18473]

Dodávateľ: ABCAM
AB190908-100 AB190908-1ML
ABCAAB190908-100EA 764.54 EUR
ABCAAB190908-100 ABCAAB190908-1ML
Anti-KAT7/Hbo1/MYST2 Rabbit Monoclonal Antibody [clone: EPR18473]
Protilátky
Rabbit monoclonal [EPR18473] to KAT7 / Hbo1 / MYST2.

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: KAT7 / Hbo1 / MYST2
Clonality: Monoclonal
Clone: EPR18473
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR